Orders LinkContactUsLinkComponent
Wipe down table
Wipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic bin, 1200×600×920 mm
  134-0489
 :   Palbam
Wipe down table
  134-0489
 :  Palbam
Wipe down table
Wipe down table
Wipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic bin, 1200×600×920 mm
1 * 1 ST
The wipe down table is designed to support the wipe down procedure. Includes a hole in the table top for easy waste disposal.
The cabinet under the table and under the waste disposal hole has a removable plastic bin included inside.
In stainless steel 304 with electropolished finish.
  • Description:
    Wipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic bin
  • W×D×H:
    1200×600×920 mm