Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides, 2
Catalog # 630-1612
Supplier:   MARIENFELD
Cavity slides
Catalog # 630-1612
Supplier:  MARIENFELD
Cavity slides
Cavity slides
Cavity slides, 2
Price
120,87 €
Log in for availability
1 * 50 ST
Product Details & Documents
Soda-lime glass of hydrolytic class 3 cavity slides.
Carefully ground and polished for in vitro diagnostic (IVD) applications.
Certifications: IVD 98/79/EC
Documents
Specifications
  • No. of wells:
    2
  • Description:
    Cavity slides
  • Seq:
    03