Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Acetylacetone 99+%

Glass bottle for solids, 250 g
Catalog # 129962500.
Toxic
Flammable
Acetylacetone 99+%
Catalog # 129962500.
CAS Number:  123-54-6
Acetylacetone 99+%
Acetylacetone 99+%

Glass bottle for solids, 250 g

CAS Number:  123-54-6
Price
37,80 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  ACAC, 2,4-Pentanedione
  • CAS Number:123-54-6
  • Formula:CH₃COCH₂COCH₃
  • MDL Number:MFCD00008787
  • Molecular Weight:100.12 g/mol
  • Merck Index:13,00083
  • Boiling Pt:133…135 °C (760 mmHg)
  • Melting Pt:–23 °C
  • Density:0.975 g/cm³
  • Flash Pt:35.5 °C
  • Storage Temperature:Ambient
  • EINECS:204-634-0
  • UN:2310
  • ADR:3,III
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4510 to 1.4530 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤50 APHA
  • Specific gravity:
    (20°C) 0.972 to 0.978
  • Acidity (CH3COOH):
    ≤0.2 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications