Order Entry
Austria
Orders LinkContactUsLinkComponent
Baumé hydrometers

Baumé hydrometers

40 - 50, 0,1, 280 mm
Catalog # ALLA0100FG050/15QP
Supplier:   Alla France
Baumé hydrometers
Catalog # ALLA0100FG050/15QP
Supplier:  Alla France
Baumé hydrometers
Baumé hydrometers

40 - 50, 0,1, 280 mm

Price
52,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Baumé hydrometers calibrated at 15 °C.

Specifications
  • Length:
    280 mm
  • Seq:
    18
  • Division:
    0,1
  • Range:
    40 - 50