Orders LinkContactUsLinkComponent
Dispenser Boxes
2 chambers, Clear, 140×160×215 mm
  212-9270
Dispenser Boxes
  212-9270
Dispenser Boxes
Dispenser Boxes
2 chambers, Clear, 140×160×215 mm
279,00 €
1 * 1 ST
Acrylic, clear.
  • Easy top loading and convenient dispensing through the front flap
  • Clear front allows contents to be checked easily
  • Cover and front flap limit infiltration by dust and other particles
Ideal for storing and dispensing a wide variety of small parts, such as pipette tips, microtubes, disposable syringes, canulas, etc.
  • Colour:
    Clear
  • Description:
    2 chambers
  • Seq:
    01
  • W×D×H:
    140×160×215 mm