Orders LinkContactUsLinkComponent
Dispenser Boxes
Dispenser Boxes
3 chambers, Clear, 210×160×215 mm
Catalog # 212-9271
Dispenser Boxes
Catalog # 212-9271
Dispenser Boxes
Dispenser Boxes
3 chambers, Clear, 210×160×215 mm
Price
317,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Acrylic, clear.
  • Easy top loading and convenient dispensing through the front flap
  • Clear front allows contents to be checked easily
  • Cover and front flap limit infiltration by dust and other particles
Ideal for storing and dispensing a wide variety of small parts, such as pipette tips, microtubes, disposable syringes, canulas, etc.
Specifications
  • Colour:
    Clear
  • Description:
    3 chambers
  • Seq:
    02
  • W×D×H:
    210×160×215 mm