Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Sample cuvettes

Sample cuvette, with lid, 17,5 ml, 50 mm, Glass
  705-0575
 :   Hach
Sample cuvettes
  705-0575
 :  Hach
Sample cuvettes
Sample cuvettes

Sample cuvette, with lid, 17,5 ml, 50 mm, Glass

 
286,00 €
   
1 * 1 ST
 

Sample cuvettes for photometric measurements.

  • Available with or without lid
  • Available reusable (made from glass) or for single use (made from plastics)
 
  • Path length:
    50 mm
  • Volume:
    17,5 ml
  • Window material:
    Glass
  • Description:
    Sample cuvette, with lid
  • Seq:
    20