Orders LinkContactUsLinkComponent
Nalgene™ Screw Caps | Avantor
Nalgene™ Screw Caps
Linerless, 28-415, White, PP
Catalog # 215-1600
   Low Manufacturing Impact
Nalgene™ Screw Caps
Catalog # 215-1600
   Low Manufacturing Impact
Nalgene™ Screw Caps
Nalgene™ Screw Caps
Linerless, 28-415, White, PP
Price
270,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
PP, coloured.
  • Very good chemical resistance
  • Heat resistant up to 135 °C
  • Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifications
  • Cap Size:
    28-415
  • Colour:
    White
  • Description:
    Linerless
  • Material:
    PP
  • Seq:
    09
  • Cat. No.:
    215-1600
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact