Orders LinkContactUsLinkComponent
Staining racks
Staining racks
Rack, for 12 slides, 435×85 mm
Catalog # DELTS-004
Supplier:   DELTALAB
Staining racks
Catalog # DELTS-004
Supplier:  DELTALAB
Staining racks
Rack, for 12 slides, 435×85 mm
Price
90,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel staining racks for microscope slides with and without dividers.
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Description:
    Rack, for 12 slides, 435×85 mm
  • Seq:
    02