Orders LinkContactUsLinkComponent
Trypsinising flasks
Trypsinising flasks
500 ml
Catalog # WITG5.507.304
Supplier:   witeg Labortechnik
Trypsinising flasks
Catalog # WITG5.507.304
Supplier:  witeg Labortechnik
Trypsinising flasks
Trypsinising flasks
500 ml
Price
48,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Trypsinising flasks, graduated, designed for converting tissue fragments and homogenates into cell suspensions by trypsin digestion of connective proteins. Flutes in flask enhance vigorous agitation.
Specifications
  • Capacity:
    500 ml
  • Seq:
    02