Orders LinkContactUsLinkComponent
Screw caps for internal thread sample tubes
Screw caps for internal thread sample tubes
Screw caps with silicone washer, Yellow
Catalog # SIMPT500YOS
Supplier:   Simport Scientific
Screw caps for internal thread sample tubes
Catalog # SIMPT500YOS
Supplier:  Simport Scientific
Screw caps for internal thread sample tubes
Screw caps for internal thread sample tubes
Screw caps with silicone washer, Yellow
Price
246,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
These polypropylene screw caps are to be used with T500 series internal thread sample tubes.
  • Made of polypropylene
  • Cap with silicone washer for leakproof seal
  • Long skirt and super-fast thread design for easy seal
  • Autoclavable
Caps are available with a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The cap features a long skirt and a super-fast thread design allowing it to be removed or screwed on with a single turn.
These silicone washer caps will guarantee a positive leakproof seal at all temperatures.
Ordering information: Tubes T500 series sold separately.
Specifications
  • Colour:
    Yellow
  • Description:
    Screw caps with silicone washer