Cannulas for micro syringes, Dosys™
Replacement needles, Stainless steel, 0,8×40 mm
Catalog # SOCO370.0840
Supplier: Socorex Isba
Jump to:
Product Details & Documents
Blunt end cannulas provide precise distribution with Luer nozzle microdispensers.
Specifications
- Ø×L:0,8×40 mm
- Description:Replacement needles
- Material:Stainless steel
- Seq:004