ContactUsLinkComponent
2313 MASTER-M Handheld Refractometer Brix 0.0 - 33.0%0.2 1 * 1 items
Catalog # 635-0403
Supplier:  Atago
2313 MASTER-M Handheld Refractometer Brix 0.0 - 33.0%0.2 1 * 1 items
Catalog # 635-0403
Supplier:  Atago
Frequently Bought Together