Order Entry
Austria
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ DEA

Solid phase extraction cartridges, Bond Elut™ DEA

Bond Elut™ Jr, DEA, 60 Å, 40 µm, 1000 mg
Catalog # VARI12166046B
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ DEA
Catalog # VARI12166046B
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ DEA

Bond Elut™ Jr, DEA, 60 Å, 40 µm, 1000 mg

Price
463,42 €
Log in for availability
1 * 100 ST
Product Details & Documents

Bond Elut™ DEA bears some resemblance to Bond Elut™ NH₂ in its properties but with a slightly lower capacity as an anion exchange sorbent. DEA has a moderately non-polar character due to the alkyl side chains on the amino functionality. These groups still afford a medium level of polarity, higher than C₈ but less polar than C₂ or CN-E.

  • More polar than C₈ but less polar than C₂ or CN
  • Lower ionic capacity than NH₂
  • Alkyl side chains confer moderately non-polar chracteristics

Bond Elut™ DEA is an excellent phase for the extraction of bisphosphonates from human serum and urine. Bisphosphonates are extremely hydrophilic and structurally similar to many endogenous phosphorylated compounds, making their selective extraction from serum or urine very challenging. The unique chemistry of Bond Elut™ DEA offers excellent selectivity of this class of compounds.

Specifications
  • Particle Size:
    40 µm
  • Pore size:
    60 Å
  • Description:
    Bond Elut™ Jr
  • Bed weight:
    1000 mg
  • Phase:
    DEA
  • Carbon load:
    8,5%
  • Cat. No.:
    VARI12166046B
  • Endcapped:
    No
  • Surface area:
    500 m²/g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.