Order Entry
Austria
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PH

Solid phase extraction cartridges, Bond Elut™ PH

Straight barrel cartridge, PH, 60 Å, 40 µm, 1 ml, 50 mg
Catalog # VARI12102062
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PH
Catalog # VARI12102062
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ PH

Straight barrel cartridge, PH, 60 Å, 40 µm, 1 ml, 50 mg

Price
287,46 €
Log in for availability
1 * 100 ST
Product Details & Documents

Bond Elut™ PH is a non-polar bonded silica material which exhibits a different selectivity to alkyl or aliphatic functionalised phases such as C8 or cyclohexyl. The electron density present in the aromatic ring affords an enhancement in the retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.

  • Added selectivity compared to other non-polar sorbents
  • Enhanced retention of planar, conjugated organic molecules
  • Similar polarity to C8
Specifications
  • Particle Size:
    40 µm
  • Pore size:
    60 Å
  • Capacity:
    1 ml
  • Description:
    Straight barrel cartridge
  • Seq:
    04
  • Bed weight:
    50 mg
  • Phase:
    PH
  • Carbon load:
    10,7%
  • Cat. No.:
    VARI12102062
  • Endcapped:
    Yes
  • Surface area:
    500 m²/g