Order Entry
Austria
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PH

Solid phase extraction cartridges, Bond Elut™ PH

LRC cartridge, PH, 60 Å, 40 µm, 10 ml, 500 mg
Catalog # VARI12113031
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PH
Catalog # VARI12113031
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ PH

LRC cartridge, PH, 60 Å, 40 µm, 10 ml, 500 mg

Price
233,24 €
Log in for availability
1 * 50 ST
Product Details & Documents

Bond Elut™ PH is a non-polar bonded silica material which exhibits a different selectivity to alkyl or aliphatic functionalised phases such as C8 or cyclohexyl. The electron density present in the aromatic ring affords an enhancement in the retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.

  • Added selectivity compared to other non-polar sorbents
  • Enhanced retention of planar, conjugated organic molecules
  • Similar polarity to C8
Specifications
  • Particle Size:
    40 µm
  • Pore size:
    60 Å
  • Capacity:
    10 ml
  • Description:
    LRC cartridge
  • Seq:
    03
  • Bed weight:
    500 mg
  • Phase:
    PH
  • Carbon load:
    10,7%
  • Cat. No.:
    VARI12113031
  • Endcapped:
    Yes
  • Surface area:
    500 m²/g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.