Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Solid phase extraction cartridges, Bond Elut™ PH

Straight barrel cartridge, PH, 60 Å, 120 µm, 3 ml, 500 mg
Catalog # VARI14102032
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PH
Catalog # VARI14102032
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ PH

Straight barrel cartridge, PH, 60 Å, 120 µm, 3 ml, 500 mg

Price
227,11 €
Log in for availability
1 * 50 ST
Product Details & Documents

Bond Elut™ PH is a non-polar bonded silica material which exhibits a different selectivity to alkyl or aliphatic functionalised phases such as C8 or cyclohexyl. The electron density present in the aromatic ring affords an enhancement in the retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.

  • Added selectivity compared to other non-polar sorbents
  • Enhanced retention of planar, conjugated organic molecules
  • Similar polarity to C8
Specifications
  • Particle Size:
    120 µm
  • Pore size:
    60 Å
  • Capacity:
    3 ml
  • Description:
    Straight barrel cartridge
  • Seq:
    10
  • Bed weight:
    500 mg
  • Phase:
    PH
  • Carbon load:
    10,7%
  • Cat. No.:
    VARI14102032
  • Endcapped:
    Yes
  • Surface area:
    500 m²/g