Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Notepad, self-adhesive, TexWrite® TexNotes

TexNotes notepad, TX5820, 76×102 mm, Light blue
  818-0124
 :   Texwipe
Notepad, self-adhesive, TexWrite® TexNotes
  818-0124
 :  Texwipe
Notepad, self-adhesive, TexWrite® TexNotes
Notepad, self-adhesive, TexWrite® TexNotes

TexNotes notepad, TX5820, 76×102 mm, Light blue

 
147,38 €
   
1 * 10 ST
 

Ideal for leaving notes on documents, reports, instruments, and most cleanroom surfaces.

  • Low contamination polymeric elastomer adhesive allows TexNotes® to be attached and removed without high residue

Format: 102×76 mm

 : 72 sheets per pad, 10 pads per pack (Bag-Within-A-Bag®).

 
  • Colour:
    Light blue
  • Description:
    TexNotes notepad, TX5820
  • Dimensions:
    76×102 mm
  • Seq:
    1