Order Entry
Austria
Orders LinkContactUsLinkComponent
Process pH combination electrodes

Process pH combination electrodes

pH combination electrode, with cylinder membrane, H 7381 HD, 2 - 13, −5...+100 °C, Glass, Gel, Ceramic, 12×120 mm, Screw plug head
Artikelnr. SCOI285090077
Anbieter:   SI Analytics
Process pH combination electrodes
Artikelnr. SCOI285090077
Anbieter:  SI Analytics
Process pH combination electrodes
Process pH combination electrodes

pH combination electrode, with cylinder membrane, H 7381 HD, 2 - 13, −5...+100 °C, Glass, Gel, Ceramic, 12×120 mm, Screw plug head

Preis
306,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Connection:
    Screw plug head
  • Ø×L:
    12×120 mm
  • Type:
    H 7381 HD
  • Description:
    pH combination electrode, with cylinder membrane
  • Material:
    Glass
  • Seq:
    07
  • Diaphr.:
    Ceramic
  • Electrolyte:
    Gel
  • Temp.:
    −5...+100 °C
  • pH:
    2 - 13