Order Entry
Austria
Orders LinkContactUsLinkComponent
Polyoxyl Stearyl Ether, Reference Standards

Polyoxyl Stearyl Ether, Reference Standards

Polyoxyl 20 Stearyl Ether, USP reference standard, Vial, 1 g
Catalog # 1547368.
Supplier:   USP
Polyoxyl Stearyl Ether, Reference Standards
Catalog # 1547368.
Supplier:  USP
CAS Number:  
Polyoxyl Stearyl Ether, Reference Standards

Polyoxyl 20 Stearyl Ether, USP reference standard, Vial, 1 g

CAS Number:  
Price
299,00 €
Log in for availability
1 * 1 g
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    1 g
  • Standards:
    USP reference standard
  • Description:
    Polyoxyl 20 Stearyl Ether
  • Cat. No.:
    1547368.
  • Formula:
    C₂₀H₄₂O₂
  • Description:
    Polyoxyl 20 Stearyl Ether
  • CAS No.:
    9005-00-9

Specifications