Technical pH buffer solutions in sachets, HI 50000 series
Product Details & Documents
These technical calibration buffers produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionised water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers.
- Technical buffer solutions with certificate
- Accuracy of ±0,01 pH at 25 °C
- Colour coded label for easy pH buffer value identification
Temperature chart of the actual pH value at various temperatures are printed on each sachets. Solutions are free from any dyes that may stain the reference cell of a pH electrode.
Certifications: Traceable to NIST Standard Reference Materials (SRMs).
Delivery information: These solutions are supplied with a certificate of analysis.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Sachet
- Reference temperature:25 °C
- pH accuracy:±0,01
- Pk Info:10×20 ml
- pH value:13,00
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



