Filter funnels
500 ml, 95 mm, 4
Catalog # LENZ07195458
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Plate Ø:95 mm
- Porosity:4
- Capacity:500 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



