Order Entry
Austria
Orders LinkContactUsLinkComponent
Filter funnels

Filter funnels

125 ml, 60 mm, 4
Catalog # LENZ07195440
Supplier:   Lenz Laborglas
Filter funnels
Catalog # LENZ07195440
Supplier:  Lenz Laborglas
Filter funnels
Filter funnels

125 ml, 60 mm, 4

Price
52,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With integrated sintered glass filters disc.

Dokumente

Specifications
  • Plate Ø:
    60 mm
  • Porosity:
    4
  • Capacity:
    125 ml