Order Entry
Austria
Orders LinkContactUsLinkComponent
Filter funnels

Filter funnels

500 ml, 95 mm, 3
Catalog # LENZ07195358
Supplier:   Lenz Laborglas
Filter funnels
Catalog # LENZ07195358
Supplier:  Lenz Laborglas
Filter funnels
Filter funnels

500 ml, 95 mm, 3

Price
83,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With integrated sintered glass filters disc.

Documents

Specifications
  • Plate Ø:
    95 mm
  • Porosity:
    3
  • Capacity:
    500 ml