Filter funnels
125 ml, 60 mm, 3
Catalog # LENZ07195340
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Dokumente
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Plate Ø:60 mm
- Porosity:3
- Capacity:125 ml



