Caffeic acid
Glass bottle for solids, 5 g
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
- CAS Number:331-39-5
- Formula:C₉H₈O₄
- MDL Number:MFCD00004392
- Molecular Weight:180.16 g/mol
- Merck Index:12,01673
- Boiling Pt:366 °C (1013 hPa)
- Melting Pt:234…237 °C
- EINECS:206-361-2
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



