Order Entry
Austria
Orders LinkContactUsLinkComponent
Caffeic acid

Caffeic acid

Glass bottle for solids, 25 g
Catalog # CAYM70602-25
Supplier:   Cayman Chemical
Caffeic acid
Catalog # CAYM70602-25
Supplier:  Cayman Chemical
CAS Number:  331-39-5
Caffeic acid

Glass bottle for solids, 25 g

CAS Number:  331-39-5
Price
154,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Formula:C₉H₈O₄
  • MDL Number:MFCD00004392
  • Molecular Weight:180.16 g/mol
  • Merck Index:12,01673
  • Boiling Pt:366 °C (1013 hPa)
  • Melting Pt:234…237 °C
  • EINECS:206-361-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.