Order Entry
Austria
Orders LinkContactUsLinkComponent
Caffeic acid

Caffeic acid

Glass bottle for solids, 10 g
Catalog # CAYM70602-10
Supplier:   Cayman Chemical
Caffeic acid
Catalog # CAYM70602-10
Supplier:  Cayman Chemical
CAS Number:  331-39-5
Caffeic acid

Glass bottle for solids, 10 g

CAS Number:  331-39-5
Price
87,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Formula:C₉H₈O₄
  • MDL Number:MFCD00004392
  • Molecular Weight:180.16 g/mol
  • Merck Index:12,01673
  • Boiling Pt:366 °C (1013 hPa)
  • Melting Pt:234…237 °C
  • EINECS:206-361-2

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications