Order Entry
Austria
Orders LinkContactUsLinkComponent
Palmitic acid

Palmitic acid

Glass bottle for solids, 50 g
Catalog # CAYM10006627-50
Supplier:   Cayman Chemical
Palmitic acid
Catalog # CAYM10006627-50
Supplier:  Cayman Chemical
CAS Number:  57-10-3
Palmitic acid

Glass bottle for solids, 50 g

CAS Number:  57-10-3
Price
253,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Saturated fatty acids are synthesised by both plants and animals from acetyl coenzyme A as a form of long-term energy storage. Palmitic acid is a common 16-carbon saturated fat that represents 10-20% of the normal human dietary fat intake, and approximately 25% of the total plasma fatty acids in plasma lipoproteins. Saturated free fatty acids induce the expression of cyclooxygenase-2. Palmitic acid methyl ester (MP) is a fatty acid ester whose concentration in cells is modulated by methanol. In studies with isolated Kupffer cells, MP inhibits phagocytosis and decreases cell viability. In cells treated with lipopolysaccharide, it also decreases secretion of interleukin-10, TNF-α, nitric oxide, and prostaglandin E2. This effect is thought to occur by the inhibition of NF-κB.

Synonyms:  Hexadecanoic acid, Hexadecylic acid
  • CAS Number:57-10-3
  • Formula:C₁₆H₃₂O₂
  • MDL Number:MFCD00002747
  • Molecular Weight:256.43 g/mol
  • Merck Index:13,07064
  • Boiling Pt:352 °C (1013 hPa)
  • Melting Pt:61…63 °C
  • Density:0.853 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-312-9

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications