Order Entry
Austria
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Micromesh™ M507

Biopsy processing/embedding cassettes, Micromesh™ M507

White
  720-0971
Biopsy processing/embedding cassettes, Micromesh™ M507
  720-0971
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507

White

 
518,00 €
   
1 * 1.000 ST
 

Acetal polymer. Biopsy processing/embedding cassettes, Micromesh™ M507.

  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems

Documents

 
  • Colour:
    White
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.