Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Tissue processing/embedding cassettes, Histosette® I, M498

Pink
Catalog # 720-0933
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M498
Catalog # 720-0933
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M498
Tissue processing/embedding cassettes, Histosette® I, M498

Pink

Price
229,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

Acetal polymer.

  • Resistant to histological solvents
  • Flow-through slots maximise fluid exchange and ensure proper drainage
  • Integral lid
  • 45° angle writing area to enable use with some automated cassette-labelling instruments
Specifications
  • Colour:
    Pink