Tissue processing/embedding cassettes, Histosette® I, M490
Beige
Catalog # 720-0893
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M490
Catalog # 720-0893
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer.
- Resistant to histological solvents
- Flow-through slots maximise fluid exchange and ensure proper drainage
- Integral lid eliminates the need for separate steel lids
- 30° writing angle
Documents
SIMPM490-11_TDS_EN SIMPM490-4_TDS_EN
Technisches Datenblatt
Specifications
- Colour:Beige
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



