Tissue processing/embedding cassettes, Histosette® I, M490
Orange
Catalog # 720-0885
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M490
Catalog # 720-0885
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer.
- Resistant to histological solvents
- Flow-through slots maximise fluid exchange and ensure proper drainage
- Integral lid eliminates the need for separate steel lids
- 30° writing angle
Documents
SIMPM490-11_TDS_EN SIMPM490-4_TDS_EN
Technisches Datenblatt
Specifications
- Colour:Orange



