Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 25, excitation TBP 400 + 495 + 570, beam splitter TFT 410 + 505 + 585, emission TBP 460 + 530 + 625
Catalog # ZEIS4880250000000
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4880250000000
Supplier:  ZEISS
Filter sets
Filter set 25, excitation TBP 400 + 495 + 570, beam splitter TFT 410 + 505 + 585, emission TBP 460 + 530 + 625
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents
Specifications
  • Description:
    Filter set 25, excitation TBP 400 + 495 + 570, beam splitter TFT 410 + 505 + 585, emission TBP 460 + 530 + 625
  • Seq:
    06