Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 43, excitation BP 545/25, beam splitter FT 570, emission BP 605/70
Catalog # ZEIS0000001114101
Supplier:   ZEISS
Filter sets
Catalog # ZEIS0000001114101
Supplier:  ZEISS
Filter sets
Filter set 43, excitation BP 545/25, beam splitter FT 570, emission BP 605/70
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents
Specifications
  • Description:
    Filter set 43, excitation BP 545/25, beam splitter FT 570, emission BP 605/70
  • Seq:
    10