Orders LinkContactUsLinkComponent
Connector kits, SeCure® 'Y'
Connector kits, SeCure® 'Y'
Connector kit for 0,18/0,25/0,28 mm column ID
Catalog # REST20276
Supplier:   Restek
Connector kits, SeCure® 'Y'
Catalog # REST20276
Supplier:  Restek
Connector kits, SeCure® 'Y'
Connector kits, SeCure® 'Y'
Connector kit for 0,18/0,25/0,28 mm column ID
Price
421,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Connector kits connects two analytical columns to a transfer line or guard column. They have a reliable seal integrity and will not unexpectedly disconnect during temperature programmed analyses. The open design allows visual confirmation of the seal for added confidence in the connection.
Combine the 'Y' Press-Tight® connector with the strength of a metal union. The ferrules and knurled nuts hold the fused silica tubing in place, which prevents the tubing from unexpectedly disconnecting, even at temperatures as high as 400 °C.
Delivery information: Kits include: SeCure® 'Y' connector body, three knurled nuts, universal 'Y' Press-Tight® union and three ferrules.
Specifications
  • Description:
    Connector kit for 0,18/0,25/0,28 mm column ID
  • Seq:
    01