Orders LinkContactUsLinkComponent
Solid phase extraction disks, Resprep®
Solid phase extraction disks, Resprep®
Solid phase extraction disks, Resprep® C8, 47 mm
Catalog # REST24048
Supplier:   Restek
Solid phase extraction disks, Resprep®
Catalog # REST24048
Supplier:  Restek
Solid phase extraction disks, Resprep®
Solid phase extraction disks, Resprep®
Solid phase extraction disks, Resprep® C8, 47 mm
Price
330,56 €
Log in for availability
1 * 24 ST
Product Details & Documents
These glass fiber disks are embedded with C18 or C8 bonded silica. Deep-pore design reduces clogging and allows faster flow rates. Semivolatile organic compounds can be etracted.
Zertifizierung: Meet the requirements for EPA methods 525.1, 506, 550.1 and 549.1.
Specifications
  • Ø:
    47 mm
  • Description:
    Solid phase extraction disks, Resprep® C8
  • Seq:
    01