Solid phase extraction cartridges, Resprep®
SPE silica cartridges, Cartridge, 6 ml, 1000 mg
Catalog # REST24038
Supplier: Restek
Jump to:
Product Details & Documents
These cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted. Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
Cartridges may be processed by any one or all of these techniques: positive pressure, sidearm flask, centrifuge, or vacuum manifold.
Specifications
- Column Format:Cartridge
- Capacity:6 ml
- Description:SPE silica cartridges
- Seq:06
- Bed weight:1000 mg