Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Resprep®
Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, with PTFE frits, Cartridge, 3 ml, 500 mg
Catalog # REST24032
Supplier:   Restek
Solid phase extraction cartridges, Resprep®
Catalog # REST24032
Supplier:  Restek
Solid phase extraction cartridges, Resprep®
Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, with PTFE frits, Cartridge, 3 ml, 500 mg
Price
Inquire For Price
Log in for availability
1 * 50 ST
Product Details & Documents
These cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted. Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
Cartridges may be processed by any one or all of these techniques: positive pressure, sidearm flask, centrifuge, or vacuum manifold.
Specifications
  • Column Format:
    Cartridge
  • Capacity:
    3 ml
  • Description:
    SPE Florisil cartridges, with PTFE frits
  • Seq:
    02
  • Bed weight:
    500 mg