Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, Cartridge, 3 ml, 500 mg
Catalog # REST24031
Supplier: Restek
Jump to:
Product Details & Documents
These cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted. Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
Cartridges may be processed by any one or all of these techniques: positive pressure, sidearm flask, centrifuge, or vacuum manifold.
Specifications
- Column Format:Cartridge
- Capacity:3 ml
- Description:SPE Florisil cartridges
- Seq:01
- Bed weight:500 mg