Orders LinkContactUsLinkComponent
Protocol wall mounts for gas regulators
Protocol wall mounts for gas regulators
Protocol station, chrome-plated brass, CGA 580 (N2, He, Ar) fitting
Catalog # REST21347
Supplier:   Restek
Protocol wall mounts for gas regulators
Catalog # REST21347
Supplier:  Restek
Protocol wall mounts for gas regulators
Protocol wall mounts for gas regulators
Protocol station, chrome-plated brass, CGA 580 (N2, He, Ar) fitting
Price
864,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
  • Ease of use
  • Prevents gas regulator damage and improves safety
Either chrome-plated brass or 316 stainless steel option is complete with a 0,9m, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.
Specifications
  • Description:
    Protocol station, chrome-plated brass, CGA 580 (N2, He, Ar) fitting
  • Seq:
    02