Filter funnels
4000 ml, 179 mm, 3
Catalog # LENZ07195379
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Plate Ø:179 mm
- Porosity:3
- Capacity:4000 ml



