Order Entry
Austria
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Glass bottle for solids, 250 g
Catalog # ROTH9139.1
Marketsource
Supplier:   Roth Carl
Tween® 80 (Polysorbate)
Catalog # ROTH9139.1
Supplier:  Roth Carl
Marketsource
CAS Number:  9005-65-6
Tween® 80 (Polysorbate)

Glass bottle for solids, 250 g

CAS Number:  9005-65-6
Price
32,70 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:∼100 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:1.062 g/cm³ (20 °C)
  • Flash Pt:>149 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.