Order Entry
Austria
Orders LinkContactUsLinkComponent
Potassium hydroxide

Potassium hydroxide

Glass bottle for solids, 1 kg
Catalog # ROTHP747.2
Corrosive
Irritant
Marketsource
Supplier:   Roth Carl
Potassium hydroxide
Catalog # ROTHP747.2
Supplier:  Roth Carl
Marketsource
CAS Number:  1310-58-3
Potassium hydroxide

Glass bottle for solids, 1 kg

CAS Number:  1310-58-3
Price
48,70 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Merck Index:12,07806
  • Boiling Pt:1320 °C (1013 hPa)
  • Melting Pt:360 °C
  • Density:2.04 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1813
  • ADR:8,II
  • REACH:01-2119487136-33
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.