Order Entry
Austria
Orders LinkContactUsLinkComponent
Potassium dichromate

Potassium dichromate

Glass bottle for solids, 1 kg
Catalog # ROTHP744.2
Oxidizing
Toxic
Corrosive
Health
Environment
Marketsource
Supplier:   Roth Carl
Potassium dichromate
Catalog # ROTHP744.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7778-50-9
Potassium dichromate

Glass bottle for solids, 1 kg

CAS Number:  7778-50-9
Price
228,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7778-50-9
  • Formula:K₂Cr₂O₇
  • MDL Number:MFCD00011367
  • Molecular Weight:294.18 g/mol
  • Merck Index:12,07790
  • Boiling Pt:500 °C (1013 hPa)
  • Melting Pt:398 °C
  • Density:2.67 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-906-6
  • UN:3087
  • ADR:5.1,II
  • REACH:01-2119454792-32
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.