Order Entry
Austria
Orders LinkContactUsLinkComponent
Potassium dichromate

Potassium dichromate

Glass bottle for solids, 500 g
Catalog # ROTHP744.1
Oxidizing
Toxic
Corrosive
Health
Environment
Marketsource
Supplier:   Roth Carl
Potassium dichromate
Catalog # ROTHP744.1
Supplier:  Roth Carl
Marketsource
CAS Number:  7778-50-9
Potassium dichromate

Glass bottle for solids, 500 g

CAS Number:  7778-50-9
Price
124,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:7778-50-9
  • Formula:K₂Cr₂O₇
  • MDL Number:MFCD00011367
  • Molecular Weight:294.18 g/mol
  • Merck Index:12,07790
  • Boiling Pt:500 °C (1013 hPa)
  • Melting Pt:398 °C
  • Density:2.67 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-906-6
  • UN:3087
  • ADR:5.1,II
  • REACH:01-2119454792-32
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications