Order Entry
Austria
Orders LinkContactUsLinkComponent
2-Propanol

2-Propanol

25 L
Catalog # ROTH9866.4
Flammable
Irritant
Marketsource
Supplier:   Roth Carl
2-Propanol
Catalog # ROTH9866.4
Supplier:  Roth Carl
Marketsource
CAS Number:  67-63-0
2-Propanol
2-Propanol

25 L

CAS Number:  67-63-0
Price
343,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0.786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications

Specifications

  • Pk:
    25 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.