D-(+)-Galactose
Glass bottle for solids, 500 g
ROTH4987.2
Marketsource
: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
- :59-23-4
- :C₆H₁₂O₆
- :MFCD00151230
- :180.16 g/mol
- :13,04356
- :527 °C (1013 hPa)
- :163…169 °C
- :1.5 g/cm³ (20 °C)
- :Ambient
- :200-416-4
- Pack type:Glass bottle for solids
- Pk:500 g