Order Entry
Austria
Orders LinkContactUsLinkComponent
Formaldehyde 35% in aqueous solution

Formaldehyde 35% in aqueous solution

30 L
Catalog # ROTH4980.3
Toxic
Corrosive
Health
Marketsource
Supplier:   Roth Carl
Formaldehyde 35% in aqueous solution
Catalog # ROTH4980.3
Supplier:  Roth Carl
Marketsource
CAS Number:  50-00-0
Formaldehyde 35% in aqueous solution

30 L

CAS Number:  50-00-0
Price
151,00 €
Log in for availability
1 * 30 L
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
Specifications

Specifications

  • Pk:
    30 L

Specifications