Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Formaldehyde 35% in aqueous solution

5 L
Catalog # ROTH4980.2
Toxic
Corrosive
Health
Marketsource
Supplier:   Roth Carl
Formaldehyde 35% in aqueous solution
Catalog # ROTH4980.2
Supplier:  Roth Carl
Marketsource
CAS Number:  50-00-0
Formaldehyde 35% in aqueous solution

5 L

CAS Number:  50-00-0
Price
49,30 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
Specifications

Specifications

  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.